1,3-Dibromo-5-isopropylbenzene 96%

Supplier: Ambeed
Warning

A758802-1G A758802-25G A758802-5G
76820-336EA 14.86 USD
76820-336 76820-338 76820-340
1,3-Dibromo-5-isopropylbenzene 96%
1,3-Dibromo-5-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀Br₂
MW: 277.9837 g/mol
MDL Number: MFCD06657957
CAS Number: 62655-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR