4,6-Dimethoxypicolinic acid 95%

Supplier: Ambeed
Warning

A754881-100MG A754881-250MG
76820-328EA 129 USD
76820-328 76820-330
4,6-Dimethoxypicolinic acid 95%
4,6-Dimethoxypicolinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MDL Number: MFCD18255876
CAS Number: 90764-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR