2,4,6-Trichloropyrimidine-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A754518-1G A754518-5G A754518-100MG A754518-250MG
76820-324EA 32 USD
76820-324 76820-326 77552-306 77552-308
2,4,6-Trichloropyrimidine-5-carboxylic acid 97%
2,4,6-Trichloropyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅HCl₃N₂O₂
MDL Number: MFCD09909820
CAS Number: 93416-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR