3-(Trifluoromethyl)aniline hydrochloride 98%

Supplier: Ambeed
Warning

A750507-100G A750507-25G A750507-5G A750507-500G
76820-310EA 50 USD
76820-310 76820-312 76820-314 77884-675
3-(Trifluoromethyl)aniline hydrochloride 98%
3-(Trifluoromethyl)aniline hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇ClF₃N
MW: 197.58 g/mol
MDL Number: MFCD00035492
CAS Number: 2646-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR