2-Fluoro-5-hydroxy-4-methylbenzoic acid 98+%

Supplier: Ambeed
Warning

A747890-100MG A747890-1G A747890-250MG A747890-5G
76820-294EA 29.71 USD
76820-294 76820-296 76820-298 76820-300
2-Fluoro-5-hydroxy-4-methylbenzoic acid 98+%
2-Fluoro-5-hydroxy-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.138 g/mol
MDL Number: MFCD19440271
CAS Number: 870221-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR