(S)-(+)-Glycidyl butyrate 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Oxiran-2-ylmethyl butyrate

A747498-100G A747498-25G A747498-5G
76820-288EA 165.51 USD
76820-288 76820-290 76820-292
(S)-(+)-Glycidyl butyrate 98%
(S)-(+)-Glycidyl butyrate

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.1684 g/mol
MDL Number: MFCD00273369
CAS Number: 65031-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR