2-(Ethoxycarbonyl)-5-fluorophenylboronic acid 95%

Supplier: Ambeed
Warning

A747432-1G A747432-250MG A747432-5G
76820-282EA 57 USD
76820-282 76820-284 76820-286
2-(Ethoxycarbonyl)-5-fluorophenylboronic acid 95%
2-(Ethoxycarbonyl)-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₄
MDL Number: MFCD09878342
CAS Number: 957062-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR