Nimodipine 95%
Supplier: Ambeed
|
Warning
|
A742694-100MG
A742694-1G
A742694-1MG
A742694-250MG
A742694-25G
A742694-5G
A742694-100G
76820-252EA
6
USD
76820-252
76820-254
76820-256
76820-258
76820-260
76820-262
77674-036
Nimodipine 95%
Nimodipine
Precaución: for Research Use Only
Formula:
C₂₁H₂₆N₂O₇ |
MDL Number:
MFCD00153848 CAS Number: 66085-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...