Ethyl 5-amino-1-(pyrimidin-2-yl)-1H-pyrazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A742463-100MG A742463-1G
76820-248EA 57.29 USD
76820-248 76820-250
Ethyl 5-amino-1-(pyrimidin-2-yl)-1H-pyrazole-4-carboxylate 95%
Ethyl 5-amino-1-(pyrimidin-2-yl)-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₅O₂
MW: 233.227 g/mol
MDL Number: MFCD02691657
CAS Number: 91129-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR