2-Fluoro-5-iodophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-5-iodophenyl)boronic acid

A735690-1G A735690-5G
76820-238EA 14.86 USD
76820-238 76820-240
2-Fluoro-5-iodophenylboronic acid 98%
2-Fluoro-5-iodophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BFIO₂
MW: 265.82 g/mol
MDL Number: MFCD05664304
CAS Number: 866683-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR