3-(Chloromethyl)pyrazole hydrochloride 98%

Supplier: Ambeed
Danger

A733592-250MG A733592-25G A733592-5G
76820-236EA 43 USD
76820-236 77884-652 77884-653
3-(Chloromethyl)pyrazole hydrochloride 98%
3-(Chloromethyl)pyrazole hydrochloride

Precaución: for Research Use Only

Formula: C₄H₅ClN₂·ClH
MDL Number: MFCD08234808
CAS Number: 69658-97-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR