2-{1-[(tert-Butoxy)carbonyl]pyrrolidin-2-yl}acetic acid 95%

Supplier: Ambeed
Danger

A733164-250MG A733164-5G A733164-1G A733164-25G
76820-232EA 14 USD
76820-232 76820-234 77674-002 77674-004
2-{1-[(tert-Butoxy)carbonyl]pyrrolidin-2-yl}acetic acid 95%
2-{1-[(tert-Butoxy)carbonyl]pyrrolidin-2-yl}acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MDL Number: MFCD02179026
CAS Number: 194154-91-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR