N-tert-Butyl 3-boronobenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: 3-[N-(tert-Butyl)sulphamoyl]phenylboronic Acid

A731655-100MG A731655-1G A731655-250MG
76820-220EA 30.77 USD
76820-220 76820-222 76820-224
N-tert-Butyl 3-boronobenzenesulfonamide 98%
N-tert-Butyl 3-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₀H₁₆BNO₄S
MW: 257.1143 g/mol
MDL Number: MFCD06659842
CAS Number: 221290-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR