4-Methyl-5-nitro-1H-indazole 95%

Supplier: Ambeed
Warning

A729609-100MG A729609-1G A729609-250MG
76820-212EA 23 USD
76820-212 76820-214 76820-216
4-Methyl-5-nitro-1H-indazole 95%
4-Methyl-5-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇N₃O₂
MDL Number: MFCD03787902
CAS Number: 101420-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR