2-fluoro-4-(methoxycarbonyl)benzoic acid 95%

Supplier: Ambeed
Warning

A728491-1G A728491-250MG A728491-5G
76820-202EA 66 USD
76820-202 76820-204 76820-206
2-fluoro-4-(methoxycarbonyl)benzoic acid 95%
2-fluoro-4-(methoxycarbonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₄
MW: 198.14 g/mol
MDL Number: MFCD20640446
CAS Number: 314241-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR