4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A728477-100MG A728477-1G A728477-250MG A728477-5G
76820-194EA 10 USD
76820-194 76820-196 76820-198 76820-200
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 95%
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MDL Number: MFCD00103235
CAS Number: 287917-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR