4-(2,5-Dimethylphenylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-((2,5-Dimethylphenyl)carbamoyl)phenyl)boronic acid

A728189-1G A728189-25G A728189-5G
76820-188EA 47 USD
76820-188 76820-190 76820-192
4-(2,5-Dimethylphenylcarbamoyl)phenylboronic acid 98%
4-(2,5-Dimethylphenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₆BNO₃
MDL Number: MFCD09027214
CAS Number: 913835-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR