2-Chloro-6-fluoro-3-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A727841-1G A727841-250MG
76820-182EA 45 USD
76820-182 76820-184
2-Chloro-6-fluoro-3-methoxybenzoic acid 98%
2-Chloro-6-fluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₃
MDL Number: MFCD04115942
CAS Number: 886499-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR