2-Ethynyl-6-methylpyridine 95%

Supplier: Ambeed
Warning

A725225-1G A725225-250MG A725225-100MG
76820-172EA 120 USD
76820-172 76820-174 77884-637
2-Ethynyl-6-methylpyridine 95%
2-Ethynyl-6-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₇N
MDL Number: MFCD08703491
CAS Number: 30413-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR