3-Methylcyclobutane-1-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methylcyclobutanecarboxylic acid

A723100-100MG A723100-1G A723100-250MG A723100-5G
76820-164EA 25.47 USD
76820-164 76820-166 76820-168 77673-982
3-Methylcyclobutane-1-carboxylic acid 95%
3-Methylcyclobutane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.142 g/mol
MDL Number: MFCD01734189
CAS Number: 57252-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR