C-(1-Methyl-1H-pyrazol-4-yl)methylamine dihydrochloride 98+%
Supplier: Ambeed
|
Danger
|
Synonyms:
(1-Methyl-1H-pyrazol-4-yl)methanamine dihydrochloride
A721955-10G
A721955-25G
A721955-5G
76820-158EA
181.42
USD
76820-158
76820-160
76820-162
C-(1-Methyl-1H-pyrazol-4-yl)methylamine dihydrochloride 98+%
C-(1-Methyl-1H-pyrazol-4-yl)methylamine dihydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₁Cl₂N₃ MW: 184.07 g/mol |
MDL Number:
MFCD09879170 CAS Number: 1185299-72-2 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...