4-(Trifluoromethyl)thiophenol 98%

Supplier: Ambeed
Danger

Synonyms: 4-Mercaptobenzotrifluoride, 4-(Trifluoromethyl)benzene-1-thiol, 4-(Trifluoromethyl)benzenethiol

A721828-1G A721828-25G A721828-5G
76820-152EA 25.47 USD
76820-152 76820-154 76820-156
4-(Trifluoromethyl)thiophenol 98%
4-(Trifluoromethyl)thiophenol

Precaución: for Research Use Only

Formula: C₇H₅F₃S
MW: 178.18 g/mol
MDL Number: MFCD00042320
CAS Number: 825-83-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR