3-Bromo-5-carboxybenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Borono-5-bromobenzoic acid, 3-Bromo-5-carboxyphenylboronic acid

A720966-1G A720966-25G A720966-5G
76820-146EA 16.98 USD
76820-146 76820-148 76820-150
3-Bromo-5-carboxybenzeneboronic acid 98%
3-Bromo-5-carboxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BBrO₄
MW: 244.835 g/mol
MDL Number: MFCD08689487
CAS Number: 913835-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR