(E)-3-(Naphth-1-yl)acrylic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2E)-3-(Naphth-1-yl)prop-2-enoic acid, (E)-3-(Naphthalen-1-yl)acrylic acid, trans-3-(Naphth-1-yl)acrylic acid

A719448-25G A719448-5G
76820-138EA 251.44 USD
76820-138 76820-140
(E)-3-(Naphth-1-yl)acrylic acid 98%
(E)-3-(Naphth-1-yl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₂
MW: 198.217 g/mol
MDL Number: MFCD00014317
CAS Number: 2006-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR