4-Chloro-3,5-dimethylphenylboronic acid 96%

Supplier: Ambeed
Warning

A715982-1G A715982-250MG A715982-5G
76820-122EA 168.69 USD
76820-122 76820-124 76820-126
4-Chloro-3,5-dimethylphenylboronic acid 96%
4-Chloro-3,5-dimethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BClO₂
MW: 184.4278 g/mol
MDL Number: MFCD10566596
CAS Number: 1056475-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR