3,5-Dibromothiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A715559-1G A715559-250MG A715559-5G
76820-116EA 92.3 USD
76820-116 76820-118 76820-120
3,5-Dibromothiophene-2-carboxylic acid 98%
3,5-Dibromothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₂Br₂O₂S
MDL Number: MFCD01006744
CAS Number: 7311-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR