4-Methylcyclohexanol 98% (mixture of isomers)
Supplier: Ambeed
|
Warning
|
A712592-25G
A712592-100G
76820-110EA
14
USD
76820-110
77884-627
4-Methylcyclohexanol 98% (mixture of isomers)
4-Methylcyclohexanol
Precaución: for Research Use Only
Formula:
C₇H₁₄O |
MDL Number:
MFCD00001449 CAS Number: 589-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...