4-Fluoro-2-methyl-1H-indole 95%

Supplier: Ambeed
Warning

A709984-100MG A709984-250MG A709984-1G
76820-096EA 92.3 USD
76820-096 76820-098 76844-910
4-Fluoro-2-methyl-1H-indole 95%
4-Fluoro-2-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈FN
MW: 149.165 g/mol
MDL Number: MFCD18380834
CAS Number: 1260383-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR