3-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Boronophenyl)-5-methyl-1,3,4-oxadiazole, (3-(5-Methyl-1,3,4-oxadiazol-2-yl)phenyl)boronic acid

A707819-100MG A707819-1G A707819-250MG
76820-086EA 37.14 USD
76820-086 76820-088 76820-090
3-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid 98%
3-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₉BN₂O₃
MW: 203.99 g/mol
MDL Number: MFCD08689540
CAS Number: 913836-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR