(3-Bromo-5-isopropylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-isopropylphenylboronic acid

A707410-1G A707410-250MG A707410-5G
76820-076EA 132.62 USD
76820-076 76820-078 76820-080
(3-Bromo-5-isopropylphenyl)boronic acid 97%
(3-Bromo-5-isopropylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BBrO₂
MW: 242.91 g/mol
MDL Number: MFCD16295265
CAS Number: 1451390-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR