2-(Boc-amino)ethanethiol 98%

Supplier: Ambeed
Danger

A705834-1G A705834-25G A705834-5G A705834-100G A705834-500G
76820-058EA 18 USD
76820-058 76820-060 76820-062 77673-928 77884-618
2-(Boc-amino)ethanethiol 98%
2-(Boc-amino)ethanethiol

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂S
MDL Number: MFCD00274335
CAS Number: 67385-09-5
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR