(2-Hydroxyphenyl)acetonitrile 97%

Supplier: Ambeed
Warning

A703512-250MG A703512-5G
76820-042EA 30.77 USD
76820-042 77569-340
(2-Hydroxyphenyl)acetonitrile 97%
(2-Hydroxyphenyl)acetonitrile

Precaución: For research use only.

Formula: C₈H₇NO
MW: 133.147 g/mol
MDL Number: MFCD00128130
CAS Number: 14714-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR