2-Chloro-5-(trifluoromethyl)benzyl alcohol 95%

Supplier: Ambeed
Warning

A703412-100G A703412-25G A703412-5G
76820-036EA 103 USD
76820-036 76820-038 76820-040
2-Chloro-5-(trifluoromethyl)benzyl alcohol 95%
2-Chloro-5-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O
MDL Number: MFCD01631469
CAS Number: 64372-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR