2-Fluoro-3-formylbenzonitrile 98%

Supplier: Ambeed
Warning

A699932-1G A699932-250MG
76820-022EA 177 USD
76820-022 76820-024
2-Fluoro-3-formylbenzonitrile 98%
2-Fluoro-3-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄FNO
MW: 149.12 g/mol
MDL Number: MFCD18394005
CAS Number: 1261823-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR