(R)-3,3'-Bis(anthracenyl-9-yl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98%

Supplier: Ambeed
Warning

Synonyms: (11bR)-2,6-Di(anthracen-9-yl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

A698128-100MG A698128-1G A698128-250MG A698128-50MG A698128-5G
76820-014EA 144.29 USD
76820-014 76820-016 76820-018 76820-020 77673-904
(R)-3,3'-Bis(anthracenyl-9-yl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98%
(R)-3,3'-Bis(anthracenyl-9-yl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Precaución: for Research Use Only

Formula: C₄₈H₂₉O₄P
MW: 700.715 g/mol
MDL Number: MFCD11045406
CAS Number: 361342-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR