4-Chloro-3-methyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

A695668-1G A695668-250MG
76820-010EA 74.27 USD
76820-010 76820-012
4-Chloro-3-methyl-1H-pyrazole 98%
4-Chloro-3-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₅ClN₂
MW: 116.55 g/mol
MDL Number: MFCD01693485
CAS Number: 15878-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR