3-Amino-2-bromo-6-fluoropyridine 97%

Supplier: Ambeed
Warning

A694019-1G A694019-25G A694019-500G
76819-996EA 14 USD
76819-996 76819-998 76820-000
3-Amino-2-bromo-6-fluoropyridine 97%
3-Amino-2-bromo-6-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
MDL Number: MFCD18262185
CAS Number: 1068976-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR