5-Methoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Formyl-4-methoxyphenylboronic acid pinacol ester
A690792-1G
A690792-250MG
A690792-5G
76819-990EA
44.56
USD
76819-990
76819-992
76819-994
5-Methoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 95%
5-Methoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₄H₁₉BO₄ MW: 262.11 g/mol |
MDL Number:
MFCD11044929 CAS Number: 1132669-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...