4-Amino-2,3-difluorobenzonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 4-Cyano-2,3-difluoroaniline

A686219-1G A686219-5G A686219-250MG
76819-974EA 38.2 USD
76819-974 76819-976 77673-876
4-Amino-2,3-difluorobenzonitrile 95%
4-Amino-2,3-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄F₂N₂
MW: 154.117 g/mol
MDL Number: MFCD09030642
CAS Number: 112279-71-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR