(2,4-Difluoro-3-(methoxycarbonyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Difluoro-3-methoxycarbonylphenylboronic acid

A683962-100MG A683962-1G A683962-250MG A683962-5G
76819-952EA 35 USD
76819-952 76819-954 76819-956 76819-958
(2,4-Difluoro-3-(methoxycarbonyl)phenyl)boronic acid 98%
(2,4-Difluoro-3-(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₇BF₂O₄
MW: 215.95 g/mol
MDL Number: MFCD16295260
CAS Number: 1190989-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR