Ethyl-1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-benzyl-5,5-difluoro-4-oxopiperidine-3-carboxylate

A683418-1G A683418-250MG
76819-948EA 115.64 USD
76819-948 76819-950
Ethyl-1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate 95%
Ethyl-1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₇F₂NO₃
MW: 297.3 g/mol
MDL Number: MFCD16036533
CAS Number: 1067915-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR