2-Methyl-1H-imidazole-4-carbaldehyde 98%

Supplier: Ambeed
Danger

Synonyms: 2-Methyl-1H-imidazole-5-carbaldehyde

A682735-100MG A682735-250MG A682735-1G
76819-944EA 24.41 USD
76819-944 76819-946 77569-304
2-Methyl-1H-imidazole-4-carbaldehyde 98%
2-Methyl-1H-imidazole-4-carbaldehyde

Precaución: For research use only.

Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD00173725
CAS Number: 35034-22-1
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR