4-Chloro-3-iodo-1H-pyrazolo[3,4-b]pyridine 95%

Supplier: Ambeed
Warning

A682658-100MG A682658-1G A682658-250MG A682658-5G
76819-936EA 41.38 USD
76819-936 76819-938 76819-940 76819-942
4-Chloro-3-iodo-1H-pyrazolo[3,4-b]pyridine 95%
4-Chloro-3-iodo-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClIN₃
MW: 279.46 g/mol
MDL Number: MFCD17016016
CAS Number: 949558-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR