N-tert-Butyl 4-boronobenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: 4-(tert-Butylsulphamoyl)benzeneboronic acid, (4-(N-(tert-Butyl)sulphamoyl)phenyl)boronic acid

A677214-1G A677214-250MG A677214-5G A677214-25G
76819-922EA 75 USD
76819-922 76819-924 76819-926 77884-573
N-tert-Butyl 4-boronobenzenesulfonamide 98%
N-tert-Butyl 4-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₀H₁₆BNO₄S
MDL Number: MFCD06659841
CAS Number: 208516-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR