4-Bromo-1-ethoxy-2-methylbenzene 98%

Supplier: Ambeed
Warning

A677041-1G A677041-250MG A677041-5G
76819-916EA 56 USD
76819-916 76819-918 76819-920
4-Bromo-1-ethoxy-2-methylbenzene 98%
4-Bromo-1-ethoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MDL Number: MFCD12547896
CAS Number: 79636-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR