3-Carbamoyl-4-fluorobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Aminocarbonyl)-4-fluorophenylboronic acid, (3-Carbamoyl-4-fluorophenyl)boronic acid

A674302-1G A674302-250MG A674302-5G
76819-894EA 119.89 USD
76819-894 76819-896 76819-898
3-Carbamoyl-4-fluorobenzeneboronic acid 98%
3-Carbamoyl-4-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BFNO₃
MW: 182.95 g/mol
MDL Number: MFCD08235053
CAS Number: 874219-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR