4-(2,3-Dimethylphenylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

A673679-1G A673679-250MG A673679-5G
76819-888EA 44.56 USD
76819-888 76819-890 76819-892
4-(2,3-Dimethylphenylcarbamoyl)phenylboronic acid 98%
4-(2,3-Dimethylphenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₆BNO₃
MW: 269.1 g/mol
MDL Number: MFCD09027210
CAS Number: 913835-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR