Methyl 1H-pyrrolo[3,2-c]pyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

A672010-100MG A672010-250MG
76819-876EA 56.23 USD
76819-876 76819-878
Methyl 1H-pyrrolo[3,2-c]pyridine-2-carboxylate 98%
Methyl 1H-pyrrolo[3,2-c]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
MDL Number: MFCD11617802
CAS Number: 853685-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR