4-(Methanesulfonamido)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-[4-(Methanesulphonylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N-(Methylsulphonyl)-4-(4,4,5,5-tetraMethyl-1,3,2-dioxaborolan-2-yl)benzenamine, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanesulphonamide, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-phenyl)methanesulphonamide, 4-(Methanesulphonylamino)phenylboronic acid Pinacol ester, 4-Methanesulphonylaminophenylboronic acid, pinacol ester, N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanesulphonamide, 4-(Methylsulphonamido)phenylboronic acid pinacol ester

A669502-1G A669502-25G A669502-5G
76819-868EA 27 USD
76819-868 76819-870 76819-872
4-(Methanesulfonamido)phenylboronic acid pinacol ester 98%
4-(Methanesulfonamido)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₄S
MDL Number: MFCD05663876
CAS Number: 616880-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR