4-Methoxybenzo[d]thiazole-2-carbonitrile 95%

Supplier: Ambeed
Warning

A667698-1G A667698-250MG
76819-852EA 255 USD
76819-852 76819-854
4-Methoxybenzo[d]thiazole-2-carbonitrile 95%
4-Methoxybenzo[d]thiazole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂OS
MDL Number: MFCD11217478
CAS Number: 7267-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR